Double spatulas, micro
210 mm, 7 mm
Katalogové číslo 231-0401
Dodavatel: RSG
Přejít na:
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
- Both ends have an angled spoon
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Quick and easy to clean
Specifikace
- Délka:210 mm
- Šířka čepele:7 mm