Orders LinkContactUsLinkComponent
Forceps with guide pin
Forceps with guide pin
Curved, with guide pin, 115 mm
Catalog # 232-0078
Supplier:   RSG
Forceps with guide pin
Catalog # 232-0078
Supplier:  RSG
Forceps with guide pin
Forceps with guide pin
Curved, with guide pin, 115 mm
Price
1 840,00 Kč
Log in for availability
1 * 5 KS
Product Details & Documents
Stainless steel, anti-magnetic. Particularly suitable for staining applications.
  • With curved, extra fine tips
  • Corrosion resistant
Specifications
  • Type:
    Curved, with guide pin
  • Délka:
    115 mm