Orders LinkContactUsLinkComponent
Forceps
Forceps
Curved, pointed, 120 mm
Catalog # RSGA116.137
Supplier:   RSG
Forceps
Catalog # RSGA116.137
Supplier:  RSG
Forceps
Forceps
Curved, pointed, 120 mm
Price
2 990,00 Kč
Log in for availability
1 * 5 KS
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic.
  • Very fine polished tips
Specifications
  • Type:
    Curved, pointed
  • Délka:
    120 mm