Orders LinkContactUsLinkComponent
Forceps, premium quality
Forceps, premium quality
Forceps, premium quality, Straight, blunt, 105 mm
Catalog # RSGA011.105
Supplier:   RSG
Forceps, premium quality
Catalog # RSGA011.105
Supplier:  RSG
Forceps, premium quality
Forceps, premium quality
Forceps, premium quality, Straight, blunt, 105 mm
Price
1 350,00 Kč
Log in for availability
1 * 5 KS
Product Details & Documents
Stainless steel 1,4301.
Specifications
  • Type:
    Straight, blunt
  • Popis:
    Forceps, premium quality
  • Délka:
    105 mm