Orders LinkContactUsLinkComponent
Forceps, premium quality
Forceps, premium quality
Forceps, premium quality, Straight, blunt, 145 mm
Catalog # RSGA011.145
Supplier:   RSG
Forceps, premium quality
Catalog # RSGA011.145
Supplier:  RSG
Forceps, premium quality
Forceps, premium quality
Forceps, premium quality, Straight, blunt, 145 mm
Přejít na:
Price
1 420,00 Kč
Přihlásit se pro dostupnost
1 * 5 KS
Product Details & Documents
Stainless steel 1,4301.
Specifikace
  • Type:
    Straight, blunt
  • Popis:
    Forceps, premium quality
  • Délka:
    145 mm