Orders LinkContactUsLinkComponent
Forceps, premium quality
Forceps, premium quality
Forceps, premium quality, Curved, pointed, 130 mm
Catalog # RSGA014.130
Supplier:   RSG
Forceps, premium quality
Catalog # RSGA014.130
Supplier:  RSG
Forceps, premium quality
Forceps, premium quality
Forceps, premium quality, Curved, pointed, 130 mm
Price
1 620,00 Kč
Log in for availability
1 * 5 KS
Product Details & Documents
Stainless steel 1,4301.
Specifications
  • Type:
    Curved, pointed
  • Popis:
    Forceps, premium quality
  • Délka:
    130 mm