Orders LinkContactUsLinkComponent
Forceps, premium quality
Forceps, premium quality
Forceps, premium quality, Straight, pointed, 200 mm
Catalog # RSGA013.200
Supplier:   RSG
Forceps, premium quality
Catalog # RSGA013.200
Supplier:  RSG
Forceps, premium quality
Forceps, premium quality
Forceps, premium quality, Straight, pointed, 200 mm
Price
3 110,00 Kč
Log in for availability
1 * 5 KS
Product Details & Documents
Stainless steel 1,4301.
Specifications
  • Type:
    Straight, pointed
  • Popis:
    Forceps, premium quality
  • Délka:
    200 mm