Dishes
10, Set, 90/115/120 mm, 175/217/245 mm
Katalogové číslo 216-1249
Dodavatel: RSG
Přejít na:
Product Details & Documents
Stainless steel, ¹⁸/₁₀.
- With folded edge; fast, easy cleaning
- Polished electrolytically, anti-magnetic
- Good chemical resistance, very good thermal resistance
Round multipurpose dishes for a wide range of uses.
Ordering information: Each dish set (Cat. No. 216-1249) comprises one of each 1, 2 and 3 litre dishes.
Specifikace
- Ø vněj.:175/217/245 mm
- Kapacita:Set
- Výška:90/115/120 mm
- Sekvence:10