Double spatulas, narrow
Stiff, 400 mm, 20 mm
RSGA830.400
: RSG
Double-ended, narrow spatulas made from stainless steel. Available flexible or non flexibe.
- Popis:Stiff
- Délka:400 mm
- Šířka čepele:20 mm
Double-ended, narrow spatulas made from stainless steel. Available flexible or non flexibe.