Orders LinkContactUsLinkComponent
Preparation forceps
Preparation forceps
09, Straight, fine, 145 mm
Catalog # HAMMHSC190-14
Supplier:   HAMMACHER KARL
Preparation forceps
Catalog # HAMMHSC190-14
Supplier:  HAMMACHER KARL
Preparation forceps
Preparation forceps
09, Straight, fine, 145 mm
Price
1 130,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel.
Specifications
  • Type:
    Straight, fine
  • Délka:
    145 mm
  • Sekvence:
    09