Reference electrodes
Reference electrode, low maintenance, Silamid® reference system, B 1981 HD, −5...+100 °C, Sklo, Referid®, KPG annular gap, 12×120 mm, Screw plug head
Katalogové číslo SCOI285111915
Dodavatel: SI Analytics
Přejít na:
Product Details & Documents
These reference electrodes are available with low maintenance plug heads or with fixed cables and free end.
Specifikace
- Připojení:Screw plug head
- Ø×D:12×120 mm
- Type:B 1981 HD
- Popis:Reference electrode, low maintenance, Silamid® reference system
- Materiál:Sklo
- Diafragma:KPG annular gap
- Elektrolyt:Referid®
- Tepl.:−5...+100 °C