VISOCOLOR® Colorimetric Tests, Chlorine
Product Details & Documents
Reagents for the determination of chlorine in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Documents
IFU Colorimetric Test Kit VISOCOLOR Alpha Chlorine.pdf IFU Colorimetric Test Kit VISOCOLOR ECO Chlorine 1 Free and Total.pdf IFU Colorimetric Test Kit VISOCOLOR ECO Chlorine 2.pdf IFU Colorimetric Test Kit VISOCOLOR ECO Chlorine 6.pdf IFU Colorimetric Test Kit VISOCOLOR ECO Swimming pool Chlorine pH.pdf
Specifikace
Specifikace
- Chemická metoda:DPD
- Bal.:1 KIT
- Popis:VISOCOLOR® ECO Chlorine 2, test no. 5-15; Measuring range on PF-12Plus. Range on other photometers can be different
- Rozměry:170×125×45 mm (Including packaging)
- Sekvence:2
- Typ:Sea water non compatible; Hazardous material: Yes
- Pro jednotku:PF-12Plus; PF-3 Drinking Water; Advance; PF-3 Pool; Visual
- Testů v kitu:150
- Rozsah:< 0,1 • 0,1 • 0,2 • 0,3 • 0,4 • 0,6 • 0,9 • 1,2 • 2,0 mg/L (Cl₂); 0,05 - 2,00 mg/L (Cl₂); 0,05 - 2,00 mg/L (Cl₂)
- Obsahy:Reagents for 150 tests, colour chart and accessories in a box
- Hmotnost:273 g (Including packaging)
- Cat. No.:MANA931015
- Plošina:Colorimetric test kits
- Životnost police (měsíce):18
- Model:MANA931015
- Skladovací teplota:15...25 °C
Specifikace
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



