Orders LinkContactUsLinkComponent
Accessories for empty solid phase extraction columns CHROMABOND®
Accessories for empty solid phase extraction columns CHROMABOND®
19, Adapters (PE) for PP columns (30 and 70 ml)
Catalog # MANA730566
Supplier:   MACHEREY-NAGEL
Accessories for empty solid phase extraction columns CHROMABOND®
Catalog # MANA730566
Supplier:  MACHEREY-NAGEL
Accessories for empty solid phase extraction columns CHROMABOND®
Accessories for empty solid phase extraction columns CHROMABOND®
19, Adapters (PE) for PP columns (30 and 70 ml)
Price
844,00 Kč
Log in for availability
1 * 1 KIT
Specifications
  • Popis:
    Adapters (PE) for PP columns (30 and 70 ml)
  • Sekvence:
    19