Orders LinkContactUsLinkComponent
Accessories for empty solid phase extraction columns CHROMABOND®
Accessories for empty solid phase extraction columns CHROMABOND®
10, Glass fiber frits for glass columns, 6 ml
Catalog # MANA730192
Supplier:   MACHEREY-NAGEL
Accessories for empty solid phase extraction columns CHROMABOND®
Catalog # MANA730192
Supplier:  MACHEREY-NAGEL
Accessories for empty solid phase extraction columns CHROMABOND®
Accessories for empty solid phase extraction columns CHROMABOND®
10, Glass fiber frits for glass columns, 6 ml
Price
6 910,00 Kč
Log in for availability
1 * 250 KS
Specifications
  • Popis:
    Glass fiber frits for glass columns, 6 ml
  • Sekvence:
    10