Bulk media for HPLC, POLYGOPREP
44, POLYGOPREP 60-20 C18, not endcapped, with 12% carbon content, 15 – 25 µm, 60 Å, 1.000 g
Katalogové číslo MANA711031.1000
Dodavatel: MACHEREY-NAGEL
Bulk media for HPLC, POLYGOPREP
Katalogové číslo MANA711031.1000
Dodavatel: MACHEREY-NAGEL
Číslo CAS:
Přejít na:
Product Details & Documents
Irregular silica phase for preparative HPLC.
- Range of pore sizes 60, 100, 300 and 1000 Å
- Available mean particle sizes 12 µm, 20 µm, 30 µm, 50 µm, 80 µm and 130 µm
- Different modifications
- pH stability 2 to 8
The irregular silica POLYGOPREP can be used for different preperative applications.
Certifikáty
Hledat podle čísla šaržeSpecifikace
Specifikace
- Velikost částic:15 – 25 µm
- Bal.:1.000 g
- Velikost pórů:60 Å
- Popis:POLYGOPREP 60-20 C<sub>18</sub>, not endcapped, with 12% carbon content
- Sekvence:44



