Bulk media for HPLC, POLYGOPREP
90, POLYGOPREP 1000-30 C4, endcapped, with <1% carbon content, 25 – 40 µm, 1000 Å, 1.000 g
Katalogové číslo MANA711026.1000
Dodavatel: MACHEREY-NAGEL
Bulk media for HPLC, POLYGOPREP
Katalogové číslo MANA711026.1000
Dodavatel: MACHEREY-NAGEL
Číslo CAS:
Přejít na:
Product Details & Documents
Irregular silica phase for preparative HPLC.
- Range of pore sizes 60, 100, 300 and 1000 Å
- Available mean particle sizes 12 µm, 20 µm, 30 µm, 50 µm, 80 µm and 130 µm
- Different modifications
- pH stability 2 to 8
The irregular silica POLYGOPREP can be used for different preperative applications.
Certifikáty
Hledat podle čísla šaržeSpecifikace
Specifikace
- Velikost částic:25 – 40 µm
- Bal.:1.000 g
- Velikost pórů:1000 Å
- Popis:POLYGOPREP 1000-30 C<sub>4</sub>, endcapped, with <1% carbon content
- Sekvence:90



