Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Bulk media for HPLC, POLYGOPREP

Bulk media for HPLC, POLYGOPREP

75, POLYGOPREP 60-20 C8, not endcapped, with 7% carbon content, 15 – 25 µm, 60 Å, 100 g
Katalogové číslo MANA711008.100
Dodavatel:   MACHEREY-NAGEL
Bulk media for HPLC, POLYGOPREP
Katalogové číslo MANA711008.100
Dodavatel:  MACHEREY-NAGEL
Číslo CAS:  
Bulk media for HPLC, POLYGOPREP
Bulk media for HPLC, POLYGOPREP

75, POLYGOPREP 60-20 C8, not endcapped, with 7% carbon content, 15 – 25 µm, 60 Å, 100 g

Přejít na:
Číslo CAS:  
Price
6 040,00 Kč
Přihlásit se pro dostupnost
1 * 100 g
Product Details & Documents

Irregular silica phase for preparative HPLC.

  • Range of pore sizes 60, 100, 300 and 1000 Å
  • Available mean particle sizes 12 µm, 20 µm, 30 µm, 50 µm, 80 µm and 130 µm
  • Different modifications
  • pH stability 2 to 8

The irregular silica POLYGOPREP can be used for different preperative applications.

Documents

Specifikace

Specifikace

  • Velikost částic:
    15 – 25 µm
  • Bal.:
    100 g
  • Velikost pórů:
    60 Å
  • Popis:
    POLYGOPREP 60-20 C<sub>8</sub>, not endcapped, with 7% carbon content
  • Sekvence:
    75

Specifikace

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.