Orders LinkContactUsLinkComponent
Extraction thimbles
Extraction thimbles
45, 30×100 mm, 1,5 mm
Katalogové číslo MANA645223
Dodavatel:   MACHEREY-NAGEL
Extraction thimbles
Katalogové číslo MANA645223
Dodavatel:  MACHEREY-NAGEL
Extraction thimbles
Extraction thimbles
45, 30×100 mm, 1,5 mm
Přejít na:
Price
5 720,00 Kč
Přihlásit se pro dostupnost
1 * 25 KS
Product Details & Documents
Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.
  • Fat determination in food and feed
  • Determination of binder content in asphalt
  • Analysis of soil contaminants
  • Determination of additives in rubber and elastomers
Specifikace
  • Vnitř. Ø×V:
    30×100 mm
  • Sekvence:
    45
  • Tloušťka:
    1,5 mm