Extraction thimbles
29, 38×200 mm, 1,5 mm
Dodavatel: MACHEREY-NAGEL
Přejít na:
Product Details & Documents
Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.
- Fat determination in food and feed
- Determination of binder content in asphalt
- Analysis of soil contaminants
- Determination of additives in rubber and elastomers
Specifikace
- Vnitř. Ø×V:38×200 mm
- Sekvence:29
- Tloušťka:1,5 mm