Orders LinkContactUsLinkComponent
Extraction thimbles
Extraction thimbles
19, 30× 90 mm, 1,5 mm
Catalog # MANA645021
Supplier:   MACHEREY-NAGEL
Extraction thimbles
Catalog # MANA645021
Supplier:  MACHEREY-NAGEL
Extraction thimbles
Extraction thimbles
19, 30× 90 mm, 1,5 mm
Price
3 460,00 Kč
Log in for availability
1 * 25 KS
Product Details & Documents
Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.
  • Fat determination in food and feed
  • Determination of binder content in asphalt
  • Analysis of soil contaminants
  • Determination of additives in rubber and elastomers
Specifications
  • Vnitř. Ø×V:
    30× 90 mm
  • Sekvence:
    19
  • Tloušťka:
    1,5 mm