Orders LinkContactUsLinkComponent
Močovina
4114, 5 kg
Catalog # A1049.5000
Supplier:   PanReac AppliChem
Močovina
Catalog # A1049.5000
Supplier:  PanReac AppliChem
CAS Number:  57-13-6
Močovina
Močovina
4114, 5 kg
CAS Number:  57-13-6
Price
7 580,00 Kč
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  • CAS Number:57-13-6
  • Formula:NH₂CONH₂
  • MDL Number:MFCD00008022
  • Molecular Weight:60.06
  • Merck Index:13,09935
  • Boiling Pt:197 °C (1013 hPa)
  • Melting Pt:132,7 °C
  • Density:1,34 g/cm³ (20 °C)
  •  :Density: 1,34 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-315-5
  • REACH:01-2119463277-33
Specifications
Specifications
  • Bal.:
    5 kg
  • Sekvence:
    4114
Specifications