Pycnometers for bituminous materials
04, Top attachment with ring mark, without numbering, 60
Catalog # NEUB0321-60-60
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
Product Details & Documents
Soda-lime glass or borosilicate glass 3.3.
- Two-part assembly both parts with identification number
- Cap with funnel for ease of filling
- Ring mark on cap stem
Specifications
- NZ:60
- Popis:Top attachment with ring mark, without numbering
- Sekvence:04