Forceps
Sharp, curved, 125 mm
Katalogové číslo 232-0037
Dodavatel: MURRAY S
Přejít na:
Product Details & Documents
Stainless steel. Dissecting forceps with guide pin, which helps to keep the fine tips aligned.
- Sharp, narrow flutes
- Curved
Specifikace
- Type:Sharp, curved
- Délka:125 mm