Orders LinkContactUsLinkComponent
Accessories for on-line multi-parameter systems, IQ sensor net
Accessories for on-line multi-parameter systems, IQ sensor net
03, SACIQ-150 2-pole connection cable for all IQ sensors with IP68 waterproof connection to the IQ Sensor Net system, 15 m
Katalogové číslo 706-0056
Dodavatel:   WTW ONLINE
Accessories for on-line multi-parameter systems, IQ sensor net
Katalogové číslo 706-0056
Dodavatel:  WTW ONLINE
Accessories for on-line multi-parameter systems, IQ sensor net
Accessories for on-line multi-parameter systems, IQ sensor net
03, SACIQ-150 2-pole connection cable for all IQ sensors with IP68 waterproof connection to the IQ Sensor Net system, 15 m
Přejít na:
Price
7 620,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    SACIQ-150 2-pole connection cable for all IQ sensors with IP68 waterproof connection to the IQ Sensor Net system, 15 m
  • Sekvence:
    03