Microscope adapters for micromanipulators
, Leica® DMIRE 2 a DMIRB/E/HC
Katalogové číslo 631-0214
Dodavatel: EPPENDORF
Přejít na:
Product Details & Documents
Microscope adapters for InjectMan® NI 2 and TransferMan® NK 2. The universal stand is suitable for mounting manipulators on upright or stereo microscopes, independent of the microscope tripod used.
Specifikace
- Popis:Leica® DMIRE 2 a DMIRB/E/HC