Orders LinkContactUsLinkComponent
Multi-way stopcocks, Discofix®
Three-way stopcocks, Modrá
Catalog # 720-2573
Supplier:   B. Braun
Multi-way stopcocks, Discofix®
Catalog # 720-2573
Supplier:  B. Braun
Multi-way stopcocks, Discofix®
Multi-way stopcocks, Discofix®
Three-way stopcocks, Modrá
Price
2 020,00 Kč
Log in for availability
1 * 50 KS
Product Details & Documents
Suitable for all pressure infusion systems <2 bar.
  • Continuous flow channels for accurate pressure monitoring
  • Swivel lock can be moved axially or radially for fast and secure connection
  • PVC and latex-free
Suitable for all pressure infusion systems <2 bar.
Specifications
  • Barva:
    Modrá
  • Popis:
    Three-way stopcocks