Orders LinkContactUsLinkComponent
Pipette tips
Pipette tips
35, Standard, 100 - 1000 µl, -, Blue, Kartell, Eppendorf, Gilson, Socorex, Brand, Nichiryo®, Sartorius, Hamilton, Rack
Catalog # KART9640
Supplier:   KARTELL
Pipette tips
Catalog # KART9640
Supplier:  KARTELL
Pipette tips
Pipette tips
35, Standard, 100 - 1000 µl, -, Blue, Kartell, Eppendorf, Gilson, Socorex, Brand, Nichiryo®, Sartorius, Hamilton, Rack
Price
2 940,00 Kč
Log in for availability
1 * 960 KS
Product Details & Documents
These tips are recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market.
  • High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
  • Precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
  • Tips in racks come with snap-on lid
  • Boxes containing 'macro' tips (5000 μl and 10000 μl) have a sliding horizontal cover
  • Containers and tips are autoclavable to +121 °C for 20 minutes
Specifications
  • Balené:
    Rack
  • Sterilní:
    -
  • Objem:
    100 - 1000 µl
  • Sekvence:
    35
  • Druh špičky:
    Standard
  • Barva špičky:
    Blue
  • Pro použití s:
    Kartell, Eppendorf, Gilson, Socorex, Brand, Nichiryo®, Sartorius, Hamilton