VARIO TOTAL CHLORINE DPD 25 ML 1 * 100 KS
Katalogové číslo LOVI530130
Dodavatel: Lovibond
Frequently Bought Together
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Immersion oil for microscopy, Sigma-Aldrich®
Dodavatel: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
Test Strips, nitrate, MQuant®, Supelco®
Dodavatel: Merck
Hydroxid sodný, pellets, EMSURE® p.a., Supelco®
Dodavatel: Merck
Joint grease, K.W.S.
Dodavatel: POLLATH
Test pro stanovení peroxidu, MQuant™, Supelco®
Dodavatel: Merck
Hydroxid sodný 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Dodavatel: VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
Dodavatel: VWR Chemicals
Test kits, COD cell test, Spectroquant®, Supelco®
Dodavatel: Merck
Chlornan sodný solution 5%, J.T.Baker®
Dodavatel: Avantor
VWR® Dosatest, pH indicator test strips, non bleeding
Dodavatel: VWR Chemicals
Test pro stanovení volného chloru, MQuant™, Supelco®
Dodavatel: Merck
Test strips, peroxide, Reflectoquant®, Supelco®
Dodavatel: Merck
VWR® Rota®, pH Indicator Paper, 1 to 14
Dodavatel: VWR Chemicals
Test kits, photometric, COD, LCK cuvette tests
Dodavatel: Hach