Pipette tips
38, Standard, 100 - 5000 µl, -, 150 mm, Zelená, Bulk
Dodavatel: Sartorius
Přejít na:
Product Details & Documents
These pipette tips are suitable for many pipettes on the market.
- Natural colour, premium grade PP, autoclavable
- Excellent seal design for accurate, leak-tight sealing
- Produced and packed in a dust-free environment under strictly maintained conditions
Informace pro objednávání: Available in bulk packaging, in autoclavable racks and in refills.
Upozornení: Article 613-4153 (BHIT780310) is designed for use with the Midi Plus Pipette Controller only.
Documents
Certifikáty
Hledat podle čísla šaržeBHIT780300_502001691.pdf BHIT780305_502033642.pdf BHIT790201_502057104.pdf BHIT790201_502068217.pdf BHIT791001_502027686.pdf
More
BHIT780300_TDS_EN BHIT780304_TDS_EN BHIT780308_TDS_EN BHIT783211_TDS_EN BHIT791002_TDS_EN
Technický list
More
Sartorius Pipette Tips
Produktová literatura
Specifikace
- Barevný kód:Zelená
- Balené:Bulk
- Sterilní:-
- Objem:100 - 5000 µl
- Délka:150 mm
- Sekvence:38
- Druh špičky:Standard
- Cat. No.:613-5090
- Certifikát ochrany životního prostředí:Renewable energy certificate; ISO 14001
- Environmentally Preferable:Low Manufacturing Impact