Orders LinkContactUsLinkComponent
Specific gravity cups, calibrated
03, Stainless steel, 50 ml
  SIMEVF2100
Specific gravity cups, calibrated
  SIMEVF2100
Specific gravity cups, calibrated
Specific gravity cups, calibrated
03, Stainless steel, 50 ml
11 110,00 Kč
1 * 1 KS
Specific gravity cups for determining the specific gravity or density (or weight per gallon wpg) of coatings, pastes or similar liquids.
  • Density is defined as weight per unit volume at a specified temperature
  • Available in anodised aluminium or stainless steel
Certifications: According to ISO 2811, DIN 53 217 and ASTM D 1475.
Delivery information: Supplied with a calibration certificate.
Documents
  • Objem (nom.):
    50 ml
  • Popis:
    Stainless steel
  • Sekvence:
    03