Calibration weights, cylindrical, OIML, class E2
19, 1 kg, 1,6 ± mg, Cylindrical test weight
Catalog # 611-0896
Supplier: Sartorius Balances
Jump to:
Product Details & Documents
Class E2 cyclindrical calibration weights.
Specifications
- Hmotnost:1 kg
- Sekvence:19
- Tolerance:1,6 ± mg
- Typ:Cylindrical test weight