Orders LinkContactUsLinkComponent
Replacement impeller for Magna Flex™ flasks
Replacement impeller for Magna Flex™ flasks
02, Replacement impeller for 250 ml Magna Flex® spinner flask
Katalogové číslo 734-3144
Dodavatel:   DWK Life Sciences
Replacement impeller for Magna Flex™ flasks
Katalogové číslo 734-3144
Dodavatel:  DWK Life Sciences
Replacement impeller for Magna Flex™ flasks
Replacement impeller for Magna Flex™ flasks
02, Replacement impeller for 250 ml Magna Flex® spinner flask
Přejít na:
Price
5 630,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
This unit includes a top cap with liners, glass stirring ball and connecting silicone tubing. This glass impeller assembly is designed for replacement on all Magna Flex™ style flasks.
Specifikace
  • Popis:
    Replacement impeller for 250 ml Magna Flex® spinner flask
  • Sekvence:
    02