Electrode Cables
10, Electrode cable, 2×B, with banana plug (Ø 4 mm), for connecting electrodes with Metrohm plug-in head G to measuring instrument, 1 m
Katalogové číslo METR6.2104.080
Dodavatel: METROHM
Přejít na:
Product Details & Documents
Specifikace
- Popis:Electrode cable, 2×B, with banana plug (Ø 4 mm), for connecting electrodes with Metrohm plug-in head G to measuring instrument
- Délka:1 m
- Sekvence:10