Jars, with lid
60 mm, 35 mm
Katalogové číslo HECH41050006
Dodavatel: GLASWARENFABRIK KARL HECHT
Přejít na:
Product Details & Documents
Jars, low form, made of glass, with notch and overlapping cover.
Specifikace
- Ø vněj.:60 mm
- Výška:35 mm
Jars, low form, made of glass, with notch and overlapping cover.