Double spatulas, micro
02, 100 mm, 35 mm, 3 mm
Catalog # 231-0031
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
- With round handle, one flat and one rounded end
- Straight shape
- Excellent thermal resistance, autoclavable
- Good chemical resistance
Specifications
- Délka:100 mm
- Sekvence:02
- Šířka čepele:3 mm
- Délka čepele:35 mm