Double spatulas, micro
07, 130 mm, 40 mm, 3 mm
Katalogové číslo 231-0032
Dodavatel: BOCHEM
Přejít na:
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
- With round handle, one flat and one rounded end
- Straight shape
- Excellent thermal resistance, autoclavable
- Good chemical resistance
Specifikace
- Délka:130 mm
- Sekvence:07
- Šířka čepele:3 mm
- Délka čepele:40 mm