Orders LinkContactUsLinkComponent
Hose clips, worm-threaded
Hose clips, worm-threaded
04, 16 - 27 mm
Catalog # BURK8677-1625
Supplier:   BURKLE
Hose clips, worm-threaded
Catalog # BURK8677-1625
Supplier:  BURKLE
Hose clips, worm-threaded
Hose clips, worm-threaded
04, 16 - 27 mm
Price
509,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
Hose clip sleeve made of stainless steel 1,4016, screw zinc coated, with screw thread.
Specifications
  • Upínací šířka:
    16 - 27 mm
  • Sekvence:
    04