Stand bases
2,6 kg, 160×250×8 mm
Catalog # BOCH5036
Supplier: BOCHEM
Jump to:
Product Details & Documents
Steel, varnished.
- With M10 threaded hole
- One adjustable foot
Certifications: DIN 12892
Specifications
- Š×H×V:160×250×8 mm
- Hmotnost:2,6 kg
Steel, varnished.
Certifications: DIN 12892