Orders LinkContactUsLinkComponent
Laboratory scissors, straight
Laboratory scissors, straight
03, Sharp/blunt, 130 mm
Catalog # BOCH4002
Supplier:   BOCHEM
Laboratory scissors, straight
Catalog # BOCH4002
Supplier:  BOCHEM
Laboratory scissors, straight
Laboratory scissors, straight
03, Sharp/blunt, 130 mm
Price
247,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Straight scissors made of stainless steel.
Caution: These scissors are not recommended for autoclaving.
Documents
Specifications
  • Type:
    Sharp/blunt
  • Délka:
    130 mm
  • Sekvence:
    03