Orders LinkContactUsLinkComponent
Forceps
Forceps
Curved, sharp, 200 mm
Catalog # BOCH1024R
Supplier:   BOCHEM
Forceps
Catalog # BOCH1024R
Supplier:  BOCHEM
Forceps
Forceps
Curved, sharp, 200 mm
Price
294,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel 18/10, antimagnetic.
Specifications
  • Type:
    Curved, sharp
  • Délka:
    200 mm