Support clips
Shaft, M12, for support clips, length 350 mm, O. Ø 15 mm
Katalogové číslo BOCH583
Dodavatel: BOCHEM
Přejít na:
Product Details & Documents
These support clips are coated with ceramic inside.
- For tubes and cylindrical vessels
- With fitting
Informace pro objednávání: The shafts (rods) need to be ordered separately.
Specifikace
- Popis:Shaft, M12, for support clips, length 350 mm, O. Ø 15 mm