Orders LinkContactUsLinkComponent
Dissecting scissors, OP-SPECIAL
Dissecting scissors, OP-SPECIAL
02, Curved, OP-special, extra sharp/sharp, 105 mm
Catalog # HAMML-HSB391-10
Supplier:   HAMMACHER KARL
Dissecting scissors, OP-SPECIAL
Catalog # HAMML-HSB391-10
Supplier:  HAMMACHER KARL
Dissecting scissors, OP-SPECIAL
Dissecting scissors, OP-SPECIAL
02, Curved, OP-special, extra sharp/sharp, 105 mm
Price
2 820,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Extra sharp dissection scissors for surgical operations.
Specifications
  • Type:
    Curved, OP-special, extra sharp/sharp
  • Délka:
    105 mm
  • Sekvence:
    02