REFERENCE SUBSTANCE FIBREBAG RS1 1 * 50 g
Katalogové číslo GERH10-0129
Dodavatel: GERHARDT
Frequently Bought Together
MULTI-ELEMENT STANDARD IN 75 BLANK OIL: 1 * 200 g
Dodavatel: LGC Standards PROMOCHEM
L-Phenylalanine, Reference Standard
Dodavatel: USP
VWR®, Cold Cranking Simulator (CCS) viscosity standards
Dodavatel: VWR Chemicals
RUTIN HYDRATE CERTIFIED 1 * 0,5 g
Dodavatel: EHRENSTORFER
Dichlofenthion (O-2,4-Dichlorophenyl O,O-diethyl phosphorothioate) 10 µg/ml in cyclohexane
Dodavatel: EHRENSTORFER
Accessories for dumatherm® (Gerhardt)
Dodavatel: GERHARDT
Pesticide Mixture 1
Dodavatel: EHRENSTORFER
CHLORHEXIDINEàDIACETATE 1 * 1 g
Dodavatel: Merck
Supelco® Organic Reference Standard, Paraquat dichloride hydrate
Dodavatel: Merck
Oil and grease standards, certified reference material, Supelco®
Dodavatel: Merck
Pour point standards, Supelco®
Dodavatel: Merck
NAPROXEN IMPURITY I 1 * 100 mg
Dodavatel: Merck
Sulfamethoxazole, analytical standard, Supelco®
Dodavatel: Merck
SO42- CRM SÍRANY 1G/L V H2O 30ML 1 * 30 mL
Dodavatel: ANALYTIKA