Double spatulas, micro
16, 150 mm, 40 mm, 6 mm
231-0601
: BOCHEM
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
- With round handle, one flat and one rounded end
- Straight shape
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Délka:150 mm
- Sekvence:16
- Šířka čepele:6 mm
- Délka čepele:40 mm