Orders LinkContactUsLinkComponent
Tissue forceps
Tissue forceps
Tissue forceps 1:2, 160 mm
Katalogové číslo 232-0004
Dodavatel:   BOCHEM
Tissue forceps
Katalogové číslo 232-0004
Dodavatel:  BOCHEM
Tissue forceps
Tissue forceps
Tissue forceps 1:2, 160 mm
Přejít na:
Price
140,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Stainless steel.
  • With small teeth (1:2) at the end of the blades
  • Designed for handling tissues with minimal trauma during surgery
Specifikace
  • Type:
    Tissue forceps 1:2
  • Délka:
    160 mm