Orders LinkContactUsLinkComponent
Dressing scissors
Dressing scissors
07, Blunt, 130 mm
Catalog # 231-1294
Supplier:   BOCHEM
Dressing scissors
Catalog # 231-1294
Supplier:  BOCHEM
Dressing scissors
Dressing scissors
07, Blunt, 130 mm
Price
155,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel. Multi-purpose scissors for a wide range of applications.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Designed for rough handling
Specifications
  • Type:
    Blunt
  • Délka:
    130 mm
  • Sekvence:
    07