Orders LinkContactUsLinkComponent
Dressing scissors
Dressing scissors
03, Sharp/blunt, 160 mm
Catalog # BOCH4102
Supplier:   BOCHEM
Dressing scissors
Catalog # BOCH4102
Supplier:  BOCHEM
Dressing scissors
Dressing scissors
03, Sharp/blunt, 160 mm
Price
168,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel. Multi-purpose scissors for a wide range of applications.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Designed for rough handling
Specifications
  • Type:
    Sharp/blunt
  • Délka:
    160 mm
  • Sekvence:
    03