Nalgene™ Screw Caps
17, Linerless, 20-415, Červený, PP
Catalog # 215-1251
Supplier: Thermo Fisher Scientific
Nalgene™ Screw Caps
Catalog # 215-1251
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PP, coloured.
- Very good chemical resistance
- Heat resistant up to 135 °C
- Autoclavable
Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.
Specifications
- Velikost víčka:20-415
- Barva:Červený
- Popis:Linerless
- Materiál:PP
- Sekvence:17
- Cat. No.:215-1251
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact