Slide mailers
003, Slide mailers, Průsvitná, PP, 84×100×6 mm, 3
Katalogové číslo 631-9161
Dodavatel: KARTELL
Přejít na:
Product Details & Documents
These mailers are suitable for the safe transportation of cytological and geological microscope slides.
- Separate compartment with guide rails and more space for each slide
- Flush closing
- Lightweight and flat
W×D×H: 50×100×6 mm (for 1); 67×85×6 mm (for 2); 84×100×6 mm (for 3)
Specifikace
- Počet sklíček:3
- Barva:Průsvitná
- Popis:Slide mailers
- Materiál:PP
- Sekvence:003
- Š×H×V:84×100×6 mm