Non-return valves
9,2 - 12,5 mm ID
Katalogové číslo 229-3331
Dodavatel: KARTELL
Přejít na:
Product Details & Documents
PP, translucent. Prevent pressure loss and backflow. Maximum working pressure 2 bar and minimum working pressure 0,07 bar at room temperature (20 °C).
- Excellent chemical resistance
- Excellent thermal resistance, autoclavable
- Tapered ends to ensure that tubes are held firmly
Documents
CoC Non-Return Valves.pdf
Prohlášení o shodě
TDS Non-return Valve 418.pdf TDS Non-return Valve 419.pdf
Technický list
Specifikace
- Velikost hadice/trubice:9,2 - 12,5 mm ID